Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TEL1
Entrez Gene ID
822211
Gene Name
terminal EAR1-like 1
Gene Aliases
AT3G26120, terminal EAR1-like 1
Chromosome Location
Chr.3: 9546259 - 9549453 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_113518.3 NP_189242.1 4 - 5 1679 99

View Full Product Details