Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GTE8
Entrez Gene ID
822345
Gene Name
global transcription factor group E8
Gene Aliases
AT3G27260, GLOBAL TRANSCRIPTION FACTOR GROUP E, global transcription factor group E8
Chromosome Location
Chr.3: 10067529 - 10072760 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001203056.2 NP_001189985.1 8 - 9 2128 84
NM_001338863.1 NP_001327403.1 8 - 9 2247 84
NM_001338864.1 NP_001327404.1 8 - 9 2220 84
NM_113640.2 NP_189362.1 8 - 9 2204 84

View Full Product Details