Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PAP29
Entrez Gene ID
836435
Gene Name
purple acid phosphatase 29
Gene Aliases
AT5G63140, ATPAP29, MDC12.10, MDC12_10, purple acid phosphatase 29
Chromosome Location
Chr.5: 25328119 - 25329882 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_125709.5 NP_201119.1 2 - 3 814 63
GenBank mRNA AY882861.1 - 2 - 3 696 63
BT014894.1 - 2 - 3 760 63
BT015022.1 - 2 - 3 696 63

View Full Product Details