Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CHR24
Entrez Gene ID
836516
Gene Name
chromatin remodeling 24
Gene Aliases
AT5G63950, ERCC6, MBM17.5, MBM17_5, RAD26, chromatin remodeling 24
Chromosome Location
Chr.5: 25591956 - 25598671 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001345616.1 NP_001332133.1 20 - 21 3129 97
NM_125791.3 NP_201200.2 20 - 21 3231 97
GenBank mRNA AF462829.1 - 20 - 21 3065 97
AY142010.1 - 20 - 21 2965 97

View Full Product Details