Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FKBP12
Entrez Gene ID
836556
Gene Name
FK506-binding protein 12
Gene Aliases
ARABIDOPSIS THALIANA FK506-BINDING PROTEIN 12, AT5G64350, ATFKBP12, FK506-binding protein 12, FKP12, IMMUNOPHILIN, MSJ1.19, MSJ1_19
Chromosome Location
Chr.5: 25734590 - 25736145 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_125831.3 NP_201240.1 1 - 2 245 72
GenBank mRNA AY059928.1 - 1 - 2 148 72
AY081564.1 - 1 - 2 90 72
U96924.1 - 1 - 2 125 72

View Full Product Details