Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FAAH
Entrez Gene ID
836565
Gene Name
fatty acid amide hydrolase
Gene Aliases
AT5G64440, AtFAAH, fatty acid amide hydrolase
Chromosome Location
Chr.5: 25765712 - 25770618 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_125840.4 NP_201249.2 11 - 12 1258 99
GenBank mRNA AY308736.1 - 11 - 12 1174 99

View Full Product Details