Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATBCAT-5
Entrez Gene ID
836707
Gene Name
branched-chain amino acid aminotransferase 5 / branched-chain amino acid transaminase 5 (BCAT5)
Gene Aliases
AT5G65780, MPA24.13, MPA24_13
Chromosome Location
Chr.5: 26311311 - 26318857 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_125975.8 NP_201379.2 6 - 7 1427 104
GenBank mRNA AJ293804.1 - 4 - 5 1139 104
AK228603.1 - 4 - 5 598 104

View Full Product Details