Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MLO10
Entrez Gene ID
836727
Gene Name
Seven transmembrane MLO family protein
Gene Aliases
AT5G65970, ATMLO10, K2A18.3, K2A18_3, MILDEW RESISTANCE LOCUS O 10
Chromosome Location
Chr.5: 26387428 - 26390767 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001345729.1 NP_001331804.1 2 - 3 527 71
NM_125994.2 NP_201398.1 2 - 3 618 71
GenBank mRNA AF369571.1 - 2 - 3 303 71
BT033095.1 - 2 - 3 258 71

View Full Product Details