Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATSAC1B
Entrez Gene ID
836732
Gene Name
Phosphoinositide phosphatase family protein
Gene Aliases
AT5G66020, ATSAC6, IBS2, IMPAIRED IN BABA-INDUCED STERILITY 2, K2A18.9, K2A18_9, SAC DOMAIN-CONTAINING PROTEIN 6, SACIB, SUPPRESSOR OF ACTIN 1B
Chromosome Location
Chr.5: 26401193 - 26404725 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_125999.3 NP_201403.2 16 - 17 1427 82
GenBank mRNA AF266458.1 - 16 - 17 1393 82
AY227249.1 - 16 - 17 1393 82

View Full Product Details