Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ADT1
Entrez Gene ID
837725
Gene Name
arogenate dehydratase 1
Gene Aliases
AT1G11790, Arabidopsis thaliana arogenate dehydratase 1, AtADT1, F25C20.4, F25C20_4, arogenate dehydratase 1
Chromosome Location
Chr.1: 3981153 - 3985219 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035947.1 NP_001031024.1 5 - 6 933 112
NM_101051.3 NP_172644.1 5 - 6 1028 112
GenBank mRNA DQ411466.1 - 5 - 6 709 112

View Full Product Details