Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
APA1
Entrez Gene ID
837740
Gene Name
aspartic proteinase A1
Gene Aliases
AT1G11910, ATAPA1, F12F1.24, F12F1_24, aspartic proteinase A1
Chromosome Location
Chr.1: 4016712 - 4021043 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001332010.1 NP_001322792.1 12 - 13 2140 74
NM_101062.4 NP_172655.1 12 - 13 1512 74
GenBank mRNA AY056387.1 - 12 - 13 1387 74
AY056403.1 - 12 - 13 1387 74
AY063974.1 - 12 - 13 1387 74
AY088657.1 - 12 - 13 1387 74
BT001980.1 - 11 - 12 1338 74

View Full Product Details