Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OZS1
Entrez Gene ID
837805
Gene Name
C4-dicarboxylate transporter/malic acid transport protein
Gene Aliases
AT1G12480, CARBON DIOXIDE INSENSITIVE 3, CDI3, F5O11.23, F5O11_23, OZONE-SENSITIVE 1, RADICAL-INDUCED CELL DEATH 3, RCD3, SLAC1, SLOW ANION CHANNEL-ASSOCIATED 1
Chromosome Location
Chr.1: 4257268 - 4259300 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_101120.3 NP_563909.1 2 - 3 1371 83
GenBank mRNA AF424557.1 - 2 - 3 1371 83
BT002640.1 - 2 - 3 1320 83

View Full Product Details