Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CYP87A2
Entrez Gene ID
837830
Gene Name
cytochrome P450, family 87, subfamily A, polypeptide 2
Gene Aliases
''cytochrome P450, AT1G12740, T12C24.27, T12C24_27, cytochrome P450, family 87, polypeptide 2, polypeptide 2'', subfamily A
Chromosome Location
Chr.1: 4342209 - 4344875 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198045.1 NP_001184974.1 3 - 4 862 81
NM_001332045.1 NP_001320967.1 3 - 4 844 81
NM_001332046.1 NP_001320968.1 3 - 4 844 81
NM_101144.5 NP_172734.2 3 - 4 917 81
GenBank mRNA AK176324.1 - 3 - 4 842 81
BT010888.1 - 3 - 4 664 81

View Full Product Details