Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATAN11
Entrez Gene ID
837849
Gene Name
Transducin/WD40 repeat-like superfamily protein
Gene Aliases
ANTHOCYANIN11, AT1G12910, F13K23.16, F13K23_16, LIGHT-REGULATED WD 1, LWD1
Chromosome Location
Chr.1: 4394897 - 4396291 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_101162.3 NP_172751.1 1 - 1 1250 143
GenBank mRNA AY093101.1 - 1 - 1 1178 143

View Full Product Details