Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sks8
Entrez Gene ID
838787
Gene Name
SKU5 similar 8
Gene Aliases
AT1G21850, SKU5 similar 8, T26F17.6, T26F17_6
Chromosome Location
Chr.1: 7667803 - 7670530 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_102033.1 NP_173603.1 5 - 6 968 61
GenBank mRNA DQ446271.1 - 5 - 6 968 61

View Full Product Details