Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EDS16
Entrez Gene ID
843810
Gene Name
ADC synthase superfamily protein
Gene Aliases
ARABIDOPSIS ISOCHORISMATE SYNTHASE 1, AT1G74710, ATICS1, ENHANCED DISEASE SUSCEPTIBILITY TO ERYSIPHE ORONTII 16, F25A4.31, ICS1, ISOCHORISMATE SYNTHASE, ISOCHORISMATE SYNTHASE 1, SALICYLIC ACID INDUCTION DEFICIENT 2, SID2
Chromosome Location
Chr.1: 28070295 - 28074177 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_106129.4 NP_565090.1 1 - 2 212 140
NM_202414.1 NP_974143.1 1 - 2 211 140
GenBank mRNA AF367342.1 - 1 - 2 210 140
AY056055.1 - 1 - 2 212 140
AY124864.1 - 1 - 2 116 140

View Full Product Details