Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sks18
Entrez Gene ID
843912
Gene Name
SKU5 similar 18
Gene Aliases
AT1G75790, F10A5.2, F10A5_2, SKU5 similar 18, SKU5 similar 18
Chromosome Location
Chr.1: 28454781 - 28457528 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_106229.2 NP_177707.1 7 - 8 1565 138
GenBank mRNA DQ446433.1 - 7 - 8 1425 138

View Full Product Details