Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CDKB2;1
Entrez Gene ID
843987
Gene Name
cyclin-dependent kinase B2;1
Gene Aliases
AT1G76540, CYCLIN-DEPENDENT KINASE B2;1, F14G6.14, F14G6_14, cyclin-dependent kinase B2;1
Chromosome Location
Chr.1: 28720328 - 28722550 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_106304.3 NP_177780.1 6 - 7 906 89
GenBank mRNA AB047279.1 - 6 - 7 707 89
AJ297936.1 - 6 - 7 707 89

View Full Product Details