Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EFS
Entrez Gene ID
844066
Gene Name
histone-lysine N-methyltransferase
Gene Aliases
ASH1 HOMOLOG 2, ASHH2, AT1G77300, CAROTENOID CHLOROPLAST REGULATORY1, CCR1, EARLY FLOWERING IN SHORT DAYS, LAZ2, LAZARUS 2, SDG8, SET DOMAIN GROUP 8, T14N5.15, T14N5_15
Chromosome Location
Chr.1: 29039922 - 29049454 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001084367.2 NP_001077836.1 2 - 3 2602 106
NM_106379.9 NP_177854.6 2 - 3 2754 106
GenBank mRNA EU014690.1 - 1 - 2 2467 106

View Full Product Details