Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AST56
Entrez Gene ID
844134
Gene Name
STAS domain / Sulfate transporter family
Gene Aliases
AT1G77990, SULPHATE TRANSPORTER 2;2, SULTR2;2
Chromosome Location
Chr.1: 29317767 - 29323352 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_106448.4 NP_565165.2 2 - 3 520 124

View Full Product Details