Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LUP2
Entrez Gene ID
844236
Gene Name
lupeol synthase 2
Gene Aliases
AT1G78960, ATLUP2, YUP8H12R.43, YUP8H12R_43, lupeol synthase 2
Chromosome Location
Chr.1: 29696394 - 29701439 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001334861.1 NP_001322959.1 12 - 13 1960 73
NM_001334862.1 NP_001322960.1 13 - 14 1726 73
NM_001334863.1 NP_001322961.1 12 - 13 1884 73
NM_001334864.1 NP_001322958.1 11 - 12 1454 73
NM_106545.4 NP_178017.2 13 - 14 1755 73
GenBank mRNA AY091131.1 - 13 - 14 1679 73
AY114039.1 - 12 - 13 1632 73

View Full Product Details