Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PTAC17
Entrez Gene ID
844387
Gene Name
plastid transcriptionally active 17
Gene Aliases
AT1G80480, T21F11.27, T21F11_27, plastid transcriptionally active 17
Chromosome Location
Chr.1: 30258005 - 30260732 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_106696.4 NP_178163.1 4 - 5 906 78
GenBank mRNA BT014948.1 - 4 - 5 862 78
BT015396.1 - 4 - 5 744 78

View Full Product Details