Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- CMT1
- Entrez Gene ID
-
844413
- Gene Name
- chromomethylase 1
- Gene Aliases
-
AT1G80740, CHROMOMETHYLASE 1, DMT4, DNA METHYLTRANSFERASE 4, F23A5.9, F23A5_9, chromomethylase 1
- Chromosome Location
-
Chr.1: 30342394 - 30347048 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_106722.2 |
NP_565245.1 |
12 - 13 |
1609 |
106 |
View Full Product Details