Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RGP3
Entrez Gene ID
820039
Gene Name
reversibly glycosylated polypeptide 3
Gene Aliases
AT3G08900, RGP, reversibly glycosylated polypeptide 3
Chromosome Location
Chr.3: 2708086 - 2709949 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_111724.3 NP_187502.2 1 - 2 563 94
GenBank mRNA AF034255.2 - 1 - 2 352 94
KJ138834.1 - 1 - 2 328 94

View Full Product Details