Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AT3G09100
Entrez Gene ID
820064
Gene Name
mRNA capping enzyme family protein
Gene Aliases
AT3G09100
Chromosome Location
Chr.3: 2787953 - 2793547 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_111745.3 NP_187522.2 9 - 10 1335 74
NM_202534.2 NP_974263.1 9 - 10 1350 74
GenBank mRNA AY125498.1 - 9 - 10 1329 74

View Full Product Details