Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RECA3
Entrez Gene ID
820177
Gene Name
RECA homolog 3
Gene Aliases
AT3G10140, RECA homolog 3
Chromosome Location
Chr.3: 3134873 - 3137417 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_111849.4 NP_187625.2 2 - 3 240 105
GenBank mRNA AY072877.1 - 2 - 3 129 105
AY136400.1 - 2 - 3 208 105
BT000214.1 - 2 - 3 129 105

View Full Product Details