Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- UNE9
- Entrez Gene ID
-
820221
- Gene Name
- Cytochrome P450 superfamily protein
- Gene Aliases
-
AT3G10560, CYP77A7, CYTOCHROME P450, FAMILY 77, POLYPEPTIDE 7, SUBFAMILY A, UNFERTILIZED EMBRYO SAC 9
- Chromosome Location
-
Chr.3: 3299893 - 3301704 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_111892.2 |
NP_187667.1 |
1 - 1 |
1637 |
120 |
View Full Product Details