Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RLP34
Entrez Gene ID
820272
Gene Name
receptor like protein 34
Gene Aliases
AT3G11010, AtRLP34, receptor like protein 34
Chromosome Location
Chr.3: 3450496 - 3453985 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_111938.3 NP_187712.2 1 - 2 2704 61

View Full Product Details