Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GC6
Entrez Gene ID
822375
Gene Name
golgin Putative 6
Gene Aliases
AT3G27530, MAG4, MAIGO 4, golgin candidate 6
Chromosome Location
Chr.3: 10193477 - 10200046 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_113669.3 NP_566820.1 18 - 19 1654 63

View Full Product Details