Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AAR3
Entrez Gene ID
822537
Gene Name
defective in cullin neddylation protein (DUF298)
Gene Aliases
AT3G28970, antiauxin-resistant 3
Chromosome Location
Chr.3: 10987785 - 10990016 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001338993.1 NP_001326021.1 8 - 9 746 85
NM_113818.4 NP_189539.1 8 - 9 860 85
GenBank mRNA AK118926.1 - 8 - 9 765 85
BT005690.1 - 8 - 9 626 85

View Full Product Details