Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- KIN11
- Entrez Gene ID
-
822566
- Gene Name
- SNF1 kinase homolog 11
- Gene Aliases
-
AKIN11, AT3G29160, ATKIN11, SNF1 kinase homolog 11, SNF1-RELATED PROTEIN KINASE, SNF1-RELATED PROTEIN KINASE 1.2, SNRK1.2
- Chromosome Location
-
Chr.3: 11128620 - 11132105 on Build TAIR10.1
- Gene Symbol
- AT2G15960
- Entrez Gene ID
-
816091
- Gene Name
- stress-induced protein
- Gene Aliases
-
AT2G15960, F19G14.4, F19G14_4
- Chromosome Location
-
Chr.2: 6947096 - 6947866 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK230337.1 |
- |
- |
763 |
108 |
View Full Product Details