Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RIK
Entrez Gene ID
822600
Gene Name
RS2-interacting KH protein
Gene Aliases
AT3G29390, RS2-interacting KH protein
Chromosome Location
Chr.3: 11289495 - 11294774 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001339023.1 NP_001326115.1 4 - 5 553 88
NM_113865.4 NP_566850.3 4 - 5 435 88
GenBank mRNA AY940684.1 - 4 - 5 395 88

View Full Product Details