Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MPK3
Entrez Gene ID
823706
Gene Name
mitogen-activated protein kinase 3
Gene Aliases
AT3G45640, ATMAPK3, ATMPK3, T6D9.4, mitogen-activated protein kinase 3
Chromosome Location
Chr.3: 16756571 - 16758874 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_114433.3 NP_190150.1 4 - 5 1099 137
GenBank mRNA AF386961.1 - 4 - 5 900 137
AK226663.1 - - 903 137
BT000007.1 - 4 - 5 752 137

View Full Product Details