Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ftsh7
Entrez Gene ID
823859
Gene Name
FTSH protease 7
Gene Aliases
AT3G47060, FTSH protease 7
Chromosome Location
Chr.3: 17332926 - 17336966 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_114573.3 NP_566889.1 12 - 13 2354 71
GenBank mRNA AY099737.1 - 12 - 13 2295 71
BT000368.1 - 12 - 13 2281 71

View Full Product Details