Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TIP5;1
Entrez Gene ID
823898
Gene Name
tonoplast intrinsic protein 5;1
Gene Aliases
AT3G47440, tonoplast intrinsic protein 5;1
Chromosome Location
Chr.3: 17481996 - 17483158 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_114612.4 NP_190328.1 2 - 3 447 98

View Full Product Details