Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MAG2
Entrez Gene ID
823924
Gene Name
RINT-1 / TIP-1 family
Gene Aliases
AT3G47700, maigo2
Chromosome Location
Chr.3: 17583601 - 17586707 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_114638.3 NP_190354.1 2 - 3 523 88

View Full Product Details