Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- CNGC16
- Entrez Gene ID
-
823956
- Gene Name
- cyclic nucleotide-gated channel 16
- Gene Aliases
-
AT3G48010, ATCNGC16, CYCLIC NUCLEOTIDE-GATED CHANNEL 16, cyclic nucleotide-gated channel 16
- Chromosome Location
-
Chr.3: 17721137 - 17724140 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_114670.2 |
NP_190384.1 |
4 - 5 |
1235 |
74 |
View Full Product Details