Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ALDH10A9
Entrez Gene ID
823972
Gene Name
aldehyde dehydrogenase 10A9
Gene Aliases
AT3G48170, BADH, BETAINE ALDEHYDE DEHYDROGENASE, aldehyde dehydrogenase 10A9
Chromosome Location
Chr.3: 17785854 - 17790026 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_114686.4 NP_190400.1 10 - 11 1134 73
GenBank mRNA AF370333.1 - 10 - 11 1082 73
AY062987.1 - 10 - 11 1026 73

View Full Product Details