Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PHT3;2
Entrez Gene ID
824046
Gene Name
phosphate transporter 3;2
Gene Aliases
AT3G48850, MPT2, mitochondrial phosphate transporter 2, phosphate transporter 3;2
Chromosome Location
Chr.3: 18114526 - 18116569 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_114744.4 NP_190454.1 3 - 4 1056 88
GenBank mRNA BT033111.1 - 3 - 4 907 88

View Full Product Details