Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RID3
Entrez Gene ID
824079
Gene Name
Transducin/WD40 repeat-like superfamily protein
Gene Aliases
AT3G49180, ROOT INITIATION DEFECTIVE 3
Chromosome Location
Chr.3: 18229747 - 18232004 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_114777.4 NP_190487.1 3 - 4 661 113
GenBank mRNA AK227986.1 - 3 - 4 630 113
BT004624.1 - 3 - 4 598 113

View Full Product Details