Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
iqd21
Entrez Gene ID
824087
Gene Name
IQ-domain 21
Gene Aliases
AT3G49260, IQ-domain 21
Chromosome Location
Chr.3: 18262290 - 18266021 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001203110.1 NP_001190039.1 2 - 3 278 88
NM_001339406.1 NP_001319716.1 2 - 3 163 88
NM_114785.4 NP_566917.1 2 - 3 387 88
NM_202676.1 NP_974405.1 1 - 2 321 88
GenBank mRNA AF361635.1 - 2 - 3 185 88
AY087861.1 - 2 - 3 226 88
AY484700.1 - 2 - 3 204 88
BT000602.1 - 1 - 2 77 88

View Full Product Details