Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
iqd15
Entrez Gene ID
824100
Gene Name
IQ-domain 15
Gene Aliases
AT3G49380, IQ-domain 15
Chromosome Location
Chr.3: 18311760 - 18313621 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_114798.2 NP_190507.1 3 - 4 711 153

View Full Product Details