Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SDN1
Entrez Gene ID
824172
Gene Name
small RNA degrading nuclease 1
Gene Aliases
AT3G50100, small RNA degrading nuclease 1
Chromosome Location
Chr.3: 18578306 - 18580686 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001339453.1 NP_001327187.1 9 - 10 1057 111
NM_114870.4 NP_190579.2 10 - 11 1132 111
GenBank mRNA BT030315.1 - 10 - 11 1027 111
EU682309.1 - 10 - 11 1027 111

View Full Product Details