Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SDN1
Entrez Gene ID
824172
Gene Name
small RNA degrading nuclease 1
Gene Aliases
AT3G50100, small RNA degrading nuclease 1
Chromosome Location
Chr.3: 18578306 - 18580686 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001339453.1 NP_001327187.1 10 - 11 1158 109
NM_114870.4 NP_190579.2 11 - 12 1233 109
GenBank mRNA BT030315.1 - 11 - 12 1128 109
EU682309.1 - 11 - 12 1128 109

View Full Product Details