Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LTI30
Entrez Gene ID
824261
Gene Name
dehydrin family protein
Gene Aliases
AT3G50970, LOW TEMPERATURE-INDUCED 30, XERO2
Chromosome Location
Chr.3: 18940643 - 18941611 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_114957.3 NP_190666.1 1 - 1 754 157
GenBank mRNA AY054260.1 - 1 - 1 596 157

View Full Product Details