Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ELP6
Entrez Gene ID
826600
Gene Name
elongator protein 6
Gene Aliases
AT4G10090, F28M11.10, F28M11_10, elongator protein 6
Chromosome Location
Chr.4: 6305320 - 6307552 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_117077.4 NP_567351.1 3 - 4 650 90
GenBank mRNA AK227863.1 - 3 - 4 602 90
AY088150.1 - 3 - 4 527 90
BT024488.1 - 3 - 4 515 90

View Full Product Details