Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATSPS4F
Entrez Gene ID
826603
Gene Name
Sucrose-phosphate synthase family protein
Gene Aliases
AT4G10120, F28M11.40, F28M11_40
Chromosome Location
Chr.4: 6313914 - 6320094 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036532.3 NP_001031609.1 7 - 8 1523 92
NM_001340652.1 NP_001329003.1 6 - 7 2464 92
NM_001340653.1 NP_001329002.1 5 - 6 1752 92
NM_001340654.1 NP_001329004.1 3 - 4 782 92
NM_117080.5 NP_192750.2 7 - 8 2373 92
GenBank mRNA KJ138763.1 - 6 - 7 1345 92

View Full Product Details