Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UBP9
Entrez Gene ID
826649
Gene Name
ubiquitin-specific protease 9
Gene Aliases
AT4G10570, T4F9.30, T4F9_30, ubiquitin-specific protease 9
Chromosome Location
Chr.4: 6523225 - 6528330 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_117125.4 NP_192795.3 1 - 2 430 131

View Full Product Details