Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UBP10
Entrez Gene ID
826651
Gene Name
ubiquitin-specific protease 10
Gene Aliases
AT4G10590, T4F9.50, T4F9_50, ubiquitin-specific protease 10
Chromosome Location
Chr.4: 6538406 - 6544915 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001340676.1 NP_001328409.1 11 - 12 2723 80
NM_001340677.1 NP_001328408.1 11 - 12 2723 80
NM_117127.2 NP_567363.1 10 - 11 1951 80
NM_179025.2 NP_849356.1 10 - 11 1989 80
GenBank mRNA AK316785.1 - 10 - 11 1953 80
AY054463.1 - 10 - 11 1952 80
BT010363.1 - 10 - 11 1775 80

View Full Product Details