Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BTI2
Entrez Gene ID
826724
Gene Name
VIRB2-interacting protein 2
Gene Aliases
AT4G11220, F8L21.10, F8L21_10, RTNLB2, Reticulan like protein B2, VIRB2-interacting protein 2
Chromosome Location
Chr.4: 6837911 - 6840036 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_117193.3 NP_192861.1 2 - 3 890 111
GenBank mRNA AY034901.1 - 2 - 3 512 111
AY048273.1 - 2 - 3 499 111
AY057606.1 - 2 - 3 433 111
AY086294.1 - 2 - 3 649 111
BT002368.1 - 2 - 3 432 111

View Full Product Details